Learn More
AdipoGen MTEP (5 mg)
SDP
Supplier:  AdipoGen AGCR10022M005
Encompass_Preferred
Chemical. CAS: 329205-68-7. Formula: C11H8N2S. MW: 200.3. Potent and selective antagonist for mGluR5. 5-fold higher anxiolytic activity than MPEP. Antidepressant. Neuroprotective. Produces antiparkinsonian-like effects in rats.
Catalog No. 50-205-3771