Learn More
AdipoGen MTEP
SDP
Catalog No. 502562393
Encompass_Preferred
Chemical. CAS 329205-68-7. Formula C11H8N2S. MW 200.3. Potent and selective antagonist for mGluR5. 5-fold higher anxiolytic activity than MPEP. Antidepressant. Neuroprotective. Produces antiparkinsonian-like effects in rats.
Catalog No. 50-256-2393 Supplier AdipoGen Supplier No. AGCR10022M025
Chemical. CAS 329205-68-7. Formula C11H8N2S. MW 200.3. Potent and selective antagonist for mGluR5. 5-fold higher anxiolytic activity than MPEP. Antidepressant. Neuroprotective. Produces antiparkinsonian-like effects in rats.