Learn More
Abcam MTEP hydrochloride (mM/ml), mGlu5 antagonist, 10MM_ML
Catalog No. 502705676
Encompass_Preferred
MW 236.72 Da, Purity >99%. Brain penetrant, potent, selective and non-competitive mGlu5 receptor antagonist (IC50 = 5 nM in Ca2+-flux assay; Ki = 16 nM). In vivo anxiolytic, devoid of side effects seen with MPEP and benzodiazepines. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5676 Supplier Abcam Supplier No. AB14448610MM_ML
MW 236.72 Da, Purity >99%. Brain penetrant, potent, selective and non-competitive mGlu5 receptor antagonist (IC50 = 5 nM in Ca2+-flux assay; Ki = 16 nM). In vivo anxiolytic, devoid of side effects seen with MPEP and benzodiazepines. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement