Learn More
Cayman Chemical MTEP (hydrochloride), Purity: ≥98%, 50 mg
SDP
Catalog No. 502970778
Encompass_Preferred
A negative allosteric modulator of the mGlu5a receptor subtype (Ki = 42 nM; IC50 = 110 nM); produces antidepressant, and anxiolytic effects
Catalog No. 50-297-0778 Supplier Cayman Chemical Supplier No. 1496150 MG
A negative allosteric modulator of the mGlu5a receptor subtype (Ki = 42 nM; IC50 = 110 nM); produces antidepressant, and anxiolytic effects