Learn More
Cayman Chemical MTEP (hydrochloride), Purity: ≥98%, 5 mg
SDP
Catalog No. 502970857
Encompass_Preferred
A negative allosteric modulator of the mGlu5a receptor subtype (Ki = 42 nM; IC50 = 110 nM); produces antidepressant, and anxiolytic effects
Catalog No. 50-297-0857 Supplier Cayman Chemical Supplier No. 149615 MG
A negative allosteric modulator of the mGlu5a receptor subtype (Ki = 42 nM; IC50 = 110 nM); produces antidepressant, and anxiolytic effects