Learn More
STEMCELL Technologies mTeSR™3D, Size: 1 Kit
SDP
Catalog No. NC1328697
Encompass
mTeSR™3D was developed for expansion and scale-up of undifferentiated human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells as aggregates in 3D suspension culture. The novel fed-batch workflow saves time and media. Daily feeds replenish nutrients and eliminate the need for medium exchanges on non-passaging days.mTeSR™3D is a specialised and serum-free formulation. This culture system is compatible with a range of suspension culture vessels.
Catalog No. NC1328697 Supplier STEMCELL Technologies Supplier No. 03950
mTeSR™3D was developed for expansion and scale-up of undifferentiated human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells as aggregates in 3D suspension culture. The novel fed-batch workflow saves time and media. Daily feeds replenish nutrients and eliminate the need for medium exchanges on non-passaging days.mTeSR™3D is a specialised and serum-free formulation. This culture system is compatible with a range of suspension culture vessels.