Learn More
STEMCELL Technologies mTeSR™ Plus
SDP
Catalog No. 502390433
Encompass
Enjoy weekend-free schedules and enhanced growth characteristics while maintaining cell quality with this stabilized feeder-free maintenance medium for human pluripotent stem cells (hPSCs). Manufactured under relevant cGMPs, mTeSR™ Plus supports fundamental research as well as cell therapy and investigational new drug research applications. Based on mTeSR™1 (Catalog #85850) with stabilized critical medium components, including FGF2, and enhanced pH buffering, Each lot of mTeSR™ Plus 5X Supplement is used to prepare complete mTeSR™ Plus medium and then performance-tested in a culture assay using hPSCs.This product is compatible with a variety of culture matrices, including Corning Matrigel hESC-Qualified Matrix and Vitronectin XF™ (Catalog #07180, developed and manufactured by Nucleus Biologics).
Catalog No. 50-239-0433 Supplier STEMCELL Technologies Supplier No. 1001130
Enjoy weekend-free schedules and enhanced growth characteristics while maintaining cell quality with this stabilized feeder-free maintenance medium for human pluripotent stem cells (hPSCs). Manufactured under relevant cGMPs, mTeSR™ Plus supports fundamental research as well as cell therapy and investigational new drug research applications. Based on mTeSR™1 (Catalog #85850) with stabilized critical medium components, including FGF2, and enhanced pH buffering, Each lot of mTeSR™ Plus 5X Supplement is used to prepare complete mTeSR™ Plus medium and then performance-tested in a culture assay using hPSCs.This product is compatible with a variety of culture matrices, including Corning Matrigel hESC-Qualified Matrix and Vitronectin XF™ (Catalog #07180, developed and manufactured by Nucleus Biologics).