Learn More
STEMCELL Technologies mTeSR™1
SDP
Catalog No. NC1595017
Encompass
Use this specialized, feeder-free culture medium to achieve more consistent human pluripotent stem cell cultures with homogenous, undifferentiated phenotypes. Manufactured under relevant cGMPs, mTeSR™1 ensures the highest quality and consistency for reproducible results in your fundamental research, as well as for cell therapy and investigational new drug research applications. This serum-free, complete cell culture medium is made with pre-screened raw materials to ensure batch-to-batch consistency and robust performance in feeder-free hPSC culture.
Catalog No. NC1595017 Supplier STEMCELL Technologies Supplier No. 85857
Use this specialized, feeder-free culture medium to achieve more consistent human pluripotent stem cell cultures with homogenous, undifferentiated phenotypes. Manufactured under relevant cGMPs, mTeSR™1 ensures the highest quality and consistency for reproducible results in your fundamental research, as well as for cell therapy and investigational new drug research applications. This serum-free, complete cell culture medium is made with pre-screened raw materials to ensure batch-to-batch consistency and robust performance in feeder-free hPSC culture.