Learn More
Aviva Systems Biology Corp MTF1 Antibody - C-terminal region (ARP32027_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32027_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MTF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MTF1. Alternate names: ZRF, MTF-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54427