Learn More
Aviva Systems Biology Corp MTF2 Antibody - middle region (ARP34291_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34291_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MTF2. Alternate names: M96, PCL2, RP5-976O13.1, dJ976O13.2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55328