Learn More
Aviva Systems Biology Corp MTFMT Antibody - C-terminal region (ARP49013_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49013_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTFMT. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MTFMT. Alternate names: FMT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-392