Learn More
Aviva Systems Biology Corp MTFR1 Antibody - C-terminal region (ARP62575_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62575_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTFR1. It was validated on Western Blot. . Alternate names: CHPPR, FAM54A2, KIAA0009ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-717