Learn More
Hach Company 2" CPVC Union Adapter, for 3700 Series Conductivity Sensors, No Tee
Catalog No. 501998052 Shop All Hach Company Products
Encompass
CPVC union adapter only - No Tee, for use with the 3700-series inductive/electrodeless conductivity sensors.
Catalog No. 50-199-8052 Supplier Hach Company Supplier No. MH538M3NZ
CPVC union adapter only - No Tee, for use with the 3700-series inductive/electrodeless conductivity sensors.