Learn More
Aviva Systems Biology Corp MTG1 Antibody - middle region (ARP62286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTG1. It was validated on Western Blot. . Alternate names: GTP, GTPBP7, RP11-108K14.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-647