Learn More
Biolog Inc MTGE BROTH - MTGE Broth, 10 Tubes/Pkg
SDP
Catalog No. NC0880730
Encompass
This item has a minimum qty of 15 per supplier requirements.
Anaerobic Enrichment Broth (MTGE BROTH) is an enriched non-selective medium used for the cultivation of most anaerobic bacteria and other fastidious microorganisms. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.MTGE Broth is a complex enriched non-selective medium developed at Anaerobe Systems for the cultivation and isolation of most anaerobic bacteria and other fastidious microorganisms. A protein formulation and yeast extract comprise the basal medium, which is then supplemented with vitamin K1, calf serum, and volatile short chain fatty acids as growth factors for many anaerobic bacteria. MTGE medium has been used to cultivate and recover difficult to grow organisms found in the human microbiota.
Catalog No. NC0880730 Supplier Biolog Inc Supplier No. AS778
This item has a minimum qty of 15 per supplier requirements.
Anaerobic Enrichment Broth (MTGE BROTH) is an enriched non-selective medium used for the cultivation of most anaerobic bacteria and other fastidious microorganisms. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.MTGE Broth is a complex enriched non-selective medium developed at Anaerobe Systems for the cultivation and isolation of most anaerobic bacteria and other fastidious microorganisms. A protein formulation and yeast extract comprise the basal medium, which is then supplemented with vitamin K1, calf serum, and volatile short chain fatty acids as growth factors for many anaerobic bacteria. MTGE medium has been used to cultivate and recover difficult to grow organisms found in the human microbiota.