Learn More
Aviva Systems Biology Corp MTHFS Antibody - middle region (ARP52186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTHFS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MTHFS. Alternate names: FLJ30410, HsT19268ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60696