Learn More
Aviva Systems Biology Corp MTHFSD Antibody - N-terminal region (ARP40988_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40988_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTHFSD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MTHFSD. Alternate names: FLJ12998, FLJ13893, MGC138262, MGC138264ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-014-57688