Learn More
Cayman Chemical mthyl alnolnyl Fluorophos 10mg
SDP
Catalog No. 502893971
Encompass_Preferred
An analog of MAFP, which has been widely studied as an inhibitor of phospholipases, FAAH, and as a CB receptor ligand; pharmacology of the α-linolenyl analog of MAFP has not been completely investigated
Catalog No. 50-289-3971 Supplier Cayman Chemical Supplier No. 7066210 MG
An analog of MAFP, which has been widely studied as an inhibitor of phospholipases, FAAH, and as a CB receptor ligand; pharmacology of the α-linolenyl analog of MAFP has not been completely investigated