Learn More
Cayman Chemical mthyl AracHDnyl Fluoropho 10mg
SDP
Catalog No. 502892263
Encompass_Preferred
An inhibitor of MAGL and FAAH (IC50s = 26.3 and 0.33 nM, respectively); irreversible inhibitor of cPLA2, as well as iPLA2 (IC50 = 500 nM); inhibits A23187-induced arachidonic acid release from human platelets (IC50 = 600 nM)
Catalog No. 50-289-2263 Supplier Cayman Chemical Supplier No. 7066010 MG
An inhibitor of MAGL and FAAH (IC50s = 26.3 and 0.33 nM, respectively); irreversible inhibitor of cPLA2, as well as iPLA2 (IC50 = 500 nM); inhibits A23187-induced arachidonic acid release from human platelets (IC50 = 600 nM)