Learn More
Cayman Chemical mthyl DIethyldIthIocrbmt 50mg
SDP
Catalog No. 502948485
Encompass_Preferred
An active metabolite of disulfiram; inhibits liver low Km ALDH in rats (ID50 = 15.5 mg/kg); decreases MAP and increases heart rate during ethanol challenge in rats at 20.6 mg/kg
Catalog No. 50-294-8485 Supplier Cayman Chemical Supplier No. 3437950 MG
An active metabolite of disulfiram; inhibits liver low Km ALDH in rats (ID50 = 15.5 mg/kg); decreases MAP and increases heart rate during ethanol challenge in rats at 20.6 mg/kg