Learn More
Cayman Chemical mthylatd LysIn PolycLNal 400uL
SDP
Catalog No. 502953544
Encompass_Preferred
A plant hormone formed by the conjugation of jasmonic acid and isoleucine signals through COI1-JAZ to direct the degradation of JAZ proteins
Catalog No. 50-295-3544 Supplier Cayman Chemical Supplier No. 13728400 UL
Add to Cart
Add to Cart
A plant hormone formed by the conjugation of jasmonic acid and isoleucine signals through COI1-JAZ to direct the degradation of JAZ proteins