Learn More
Cayman Chemical Methylchloroisothiazolinone/Methylisothiazolinone Mixture, CAS: 55965-84-9, 100 mL
SDP
Catalog No. 502941561
Encompass_Preferred
A mixture of isothiazolinone biocides; effective against Gram-positive and Gram-negative bacteria (MICs = 0.0002, 0.0002, 0.00005, and 0.00005% (w/w) for S. aureus, P. aeruginosa, A. niger, and C. albicans, respectively); can elicit contact sensitization
Catalog No. 50-294-1561 Supplier Cayman Chemical Supplier No. 27470100 ML
A mixture of isothiazolinone biocides; effective against Gram-positive and Gram-negative bacteria (MICs = 0.0002, 0.0002, 0.00005, and 0.00005% (w/w) for S. aureus, P. aeruginosa, A. niger, and C. albicans, respectively); can elicit contact sensitization