Learn More
TARGETMOL CHEMICALS INC MTI-31 1MG
SDP
Catalog No. 502878719
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.MTI-31 (LXI-15029) is a potent, orally active, and highly selective inhibitor of mTORC1 and mTORC2, exhibiting a Kd of 0.20 nM for mTOR and >5,000-fold selectivity over PIK3CA, PIK3CB, and PIK3G in binding assays, with an IC50 of 39 nM for mTOR in a LANCE assay of mTOR substrate phosphorylation with 100 uM ATP [1]. Purity 99.76%
Catalog No. 50-287-8719 Supplier TARGETMOL CHEMICALS INC Supplier No. T353431MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.MTI-31 (LXI-15029) is a potent, orally active, and highly selective inhibitor of mTORC1 and mTORC2, exhibiting a Kd of 0.20 nM for mTOR and >5,000-fold selectivity over PIK3CA, PIK3CB, and PIK3G in binding assays, with an IC50 of 39 nM for mTOR in a LANCE assay of mTOR substrate phosphorylation with 100 uM ATP [1]. Purity 99.76%