Learn More
Medchemexpress LLC Mti-31 (LXI-15029) | 1567915-38-1 | 100.0% | 474.55 g/mol | C26H30N6O3 | 25MG
SDP
Catalog No. 5000236438
Encompass_Preferred
MTI-31 (LXI-15029) is a potent, selective dual mTORC1/mTORC2 inhibitor used in preclinical cancer research. It exhibits high affinity for mTOR (reported Kd ≈ 0.20 nM; IC50 ≈ 39 nM) and has demonstrated inhibition of tumor growth, epithelial-mesenchymal transition, and metastasis in model systems. Supplied as a white-to-yellow solid for research use only.
  • Potent dual mTORC1/mTORC2 inhibition with subnanomolar binding affinity.
  • Demonstrated activity in preclinical tumor and metastasis models.
  • High reported purity for analytical consistency.
  • Stable solid form with recommended refrigerated storage conditions.
  • Suitable for both in vitro and in vivo research applications.
Catalog No. 50-002-36438 Supplier Medchemexpress LLC Supplier No. HY12607725MG
MTI-31 (LXI-15029) is a potent, selective dual mTORC1/mTORC2 inhibitor used in preclinical cancer research. It exhibits high affinity for mTOR (reported Kd ≈ 0.20 nM; IC50 ≈ 39 nM) and has demonstrated inhibition of tumor growth, epithelial-mesenchymal transition, and metastasis in model systems. Supplied as a white-to-yellow solid for research use only.
  • Potent dual mTORC1/mTORC2 inhibition with subnanomolar binding affinity.
  • Demonstrated activity in preclinical tumor and metastasis models.
  • High reported purity for analytical consistency.
  • Stable solid form with recommended refrigerated storage conditions.
  • Suitable for both in vitro and in vivo research applications.