Learn More
MSC Mitutoyo SD-6" AX Electronic Linear Scale, 0 to 6 in., 0.0005 in. Resolution, 1 ea.
Catalog No. 5000514835
Encompass
This horizontal electronic linear scale is a single-function, digital measuring tool designed for precise linear distance measurements. Featuring an absolute measurement function, the scale allows for accurate, repeatable readings in both metric and imperial units. It includes two mounting brackets for secure installation and is equipped with SPC data output capabilities for efficient data management.
  • Measurement range of 0 to 150 mm (0 to 6 in.)
  • Accuracy of 0.03 mm (0.001 in.)
  • Resolution of 0.01 mm (0.0005 in.)
  • Approximate battery life of 18,000 hours
  • Stainless-steel construction
  • Equipped with SPC data output
  • Includes two mounting brackets
Catalog No. 50-005-14835 Supplier MSC Supplier No. 01129675
This horizontal electronic linear scale is a single-function, digital measuring tool designed for precise linear distance measurements. Featuring an absolute measurement function, the scale allows for accurate, repeatable readings in both metric and imperial units. It includes two mounting brackets for secure installation and is equipped with SPC data output capabilities for efficient data management.
  • Measurement range of 0 to 150 mm (0 to 6 in.)
  • Accuracy of 0.03 mm (0.001 in.)
  • Resolution of 0.01 mm (0.0005 in.)
  • Approximate battery life of 18,000 hours
  • Stainless-steel construction
  • Equipped with SPC data output
  • Includes two mounting brackets