Learn More
MSC Mitutoyo Test Indicator Contact Point, 0.0394 in Ball Diameter, Steel
Catalog No. 5000516562
Encompass
This test indicator contact point is a precision accessory designed for accurate measurements. It features a specific ball diameter and is constructed from steel for durability and reliable performance.
  • Features a 0.0394 in (1.00 mm) ball diameter.
  • Constructed from durable steel for long-lasting use.
  • Compatible with 513 Series Dial Test Indicators.
Catalog No. 50-005-16562 Supplier MSC Supplier No. 09729567
This test indicator contact point is a precision accessory designed for accurate measurements. It features a specific ball diameter and is constructed from steel for durability and reliable performance.
  • Features a 0.0394 in (1.00 mm) ball diameter.
  • Constructed from durable steel for long-lasting use.
  • Compatible with 513 Series Dial Test Indicators.