Learn More
MSC Mitutoyo Radius Gage, 15.5 mm to 25 mm, 30 Leaves, 1 ea
Catalog No. 5000511186
Encompass
This radius gage set is designed for precise measurement of internal and external radii. Each leaf features a 180° arc and comprises both a concave and convex radius gage of the same size, with the radius dimensions clearly stamped on each leaf for easy identification.
  • Range of 15.5 mm to 25 mm
  • Includes 30 leaves
  • Leaves cover 15.5 mm to 20 mm by 0.5 mm and 21 mm to 25 mm by 1 mm increments
  • Features five measuring locations per gage
  • Equipped with a locking clamp for secure use
Catalog No. 50-005-11186 Supplier MSC Supplier No. 06242655
This radius gage set is designed for precise measurement of internal and external radii. Each leaf features a 180° arc and comprises both a concave and convex radius gage of the same size, with the radius dimensions clearly stamped on each leaf for easy identification.
  • Range of 15.5 mm to 25 mm
  • Includes 30 leaves
  • Leaves cover 15.5 mm to 20 mm by 0.5 mm and 21 mm to 25 mm by 1 mm increments
  • Features five measuring locations per gage
  • Equipped with a locking clamp for secure use