Learn More
MSC Mitutoyo Tubular-Rod Inside Micrometer Set, 2 to 12 in., Carbide-Tipped, 10/Set
Catalog No. 5000511179
Encompass
This tubular single-rod type inside micrometer set is designed for precise internal dimension measurements ranging from 2 in. to 12 in. The set includes 10 individual micrometer units, each featuring durable carbide-tipped measuring faces for enhanced wear resistance. The satin chrome finished micrometer head provides clear, crisp graduations to ensure accurate readings. Users can easily readjust the zero point by rotating the micrometer head sleeve, with a key wrench supplied for convenience.
  • Measurement range of 2 in. to 12 in.
  • Includes 10 micrometer units with locking clamp
  • Carbide-tipped measuring faces
  • Satin chrome finish on micrometer head for clear graduations
  • Adjustable zero point via rotating sleeve
  • Supplied in a fitted case
  • Key wrench included
Catalog No. 50-005-11179 Supplier MSC Supplier No. 06234793
This tubular single-rod type inside micrometer set is designed for precise internal dimension measurements ranging from 2 in. to 12 in. The set includes 10 individual micrometer units, each featuring durable carbide-tipped measuring faces for enhanced wear resistance. The satin chrome finished micrometer head provides clear, crisp graduations to ensure accurate readings. Users can easily readjust the zero point by rotating the micrometer head sleeve, with a key wrench supplied for convenience.
  • Measurement range of 2 in. to 12 in.
  • Includes 10 micrometer units with locking clamp
  • Carbide-tipped measuring faces
  • Satin chrome finish on micrometer head for clear graduations
  • Adjustable zero point via rotating sleeve
  • Supplied in a fitted case
  • Key wrench included