Learn More
MSC Mitutoyo Uni-Mike Interchangeable Anvil Micrometer, 0 to 25 mm, 0.01 mm Graduation, Carbide Tips, 1 ea
Catalog No. 5000516891
Encompass
This Uni-Mike micrometer is an interchangeable anvil tool designed for high-precision measurement of tube thickness, shoulder-edge distances, and rivet head heights. The mechanical device features carbide-tipped measuring faces to ensure long-term accuracy and a durable frame with a baked enamel finish.
  • Measurement range of 0 mm to 25 mm with 0.01 mm graduations
  • Accuracy of ± 0.004 mm
  • Ratchet stop mechanism provides an audible click for constant measuring force
  • Satin chrome finish on sleeve and laser-etched thimble markings for readability
  • Supplied with flat and rod anvils in a fitted plastic case
  • Certificate of inspection included
Catalog No. 50-005-16891 Supplier MSC Supplier No. 06233472
This Uni-Mike micrometer is an interchangeable anvil tool designed for high-precision measurement of tube thickness, shoulder-edge distances, and rivet head heights. The mechanical device features carbide-tipped measuring faces to ensure long-term accuracy and a durable frame with a baked enamel finish.
  • Measurement range of 0 mm to 25 mm with 0.01 mm graduations
  • Accuracy of ± 0.004 mm
  • Ratchet stop mechanism provides an audible click for constant measuring force
  • Satin chrome finish on sleeve and laser-etched thimble markings for readability
  • Supplied with flat and rod anvils in a fitted plastic case
  • Certificate of inspection included