Learn More
Aviva Systems Biology Corp MTIF3 Antibody - middle region (ARP55563_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55563_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTIF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MTIF3. Alternate names: IF-3mt, IF3(mt), IF3mtApplicationWB. Species reactivity: Human
Catalog No. 50-014-61539