Learn More
Medchemexpress LLC MTL-CEBPA sodium | 00-00-0 | 94.6% | 13,464.27 g/mol | 5 MG
SDP
Catalog No. 5000263255
Encompass_Preferred
MTL-CEBPA sodium is the sodium salt form of a small activating RNA (saRNA) designed to upregulate the transcription factor C/EBPα. Supplied as a solid for research use, it is intended for gene-activation studies in inflammation and cancer research and is formulated for low-temperature storage and handling.
  • Activates C/EBPα expression in research models.
  • Supplied as a stable solid sodium salt suitable for low-temperature storage.
  • High purity suitable for research applications.
  • Available in small milligram-scale package sizes for experimental use.
  • Storage guidance provided for solvent and solid conditions.
Catalog No. 50-002-63255 Supplier Medchemexpress LLC Supplier No. HY132607A5MG
MTL-CEBPA sodium is the sodium salt form of a small activating RNA (saRNA) designed to upregulate the transcription factor C/EBPα. Supplied as a solid for research use, it is intended for gene-activation studies in inflammation and cancer research and is formulated for low-temperature storage and handling.
  • Activates C/EBPα expression in research models.
  • Supplied as a stable solid sodium salt suitable for low-temperature storage.
  • High purity suitable for research applications.
  • Available in small milligram-scale package sizes for experimental use.
  • Storage guidance provided for solvent and solid conditions.