Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Vikan Bench Scraper, Metal Detectable, Polypropylene, 8.3 in, Blue

Catalog No. 502838983
Encompass

This metal detectable bench scraper is ideal for bench top scraping, utilizing a semi-ferrous additive for metal detection system compatibility. Its lightweight, durable polypropylene construction offers a cost-effective solution for environments prioritizing hygiene and food safety.

  • Contains semi-ferrous additives for metal/X-ray detectability.
  • Beveled and straight scraping edges for effective cleaning.
  • Durable polypropylene construction ensures long-lasting performance.
  • One-piece, injection-molded design prevents bacterial growth.
  • Easy to clean and maintain for high hygiene standards.
  • Color-coded for hygienic zoning, 5S, and HACCP to prevent cross-contamination.
  • Built-in handle with slotted grip for optimal control.
  • Will not scratch metal surfaces.
  • FDA compliant.
  • Dishwasher and autoclave safe.

Catalog No. 50-283-8983 Supplier MSC Supplier No. 42033167
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This metal detectable bench scraper is ideal for bench top scraping, utilizing a semi-ferrous additive for metal detection system compatibility. Its lightweight, durable polypropylene construction offers a cost-effective solution for environments prioritizing hygiene and food safety.

  • Contains semi-ferrous additives for metal/X-ray detectability.
  • Beveled and straight scraping edges for effective cleaning.
  • Durable polypropylene construction ensures long-lasting performance.
  • One-piece, injection-molded design prevents bacterial growth.
  • Easy to clean and maintain for high hygiene standards.
  • Color-coded for hygienic zoning, 5S, and HACCP to prevent cross-contamination.
  • Built-in handle with slotted grip for optimal control.
  • Will not scratch metal surfaces.
  • FDA compliant.
  • Dishwasher and autoclave safe.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.