Learn More
MSC Vikan Mini Bench Scraper, Metal Detectable, 5.2 in, White
Catalog No. 502838411
Encompass
This metal detectable mini bench scraper provides an excellent solution for bench top scraping needs. Its semi-ferrous additive allows for use with metal detectable systems. This plastic scraper offers a lightweight and less costly alternative to stainless steel.
  • Metal detectable
  • Semi-ferrous additive for use with metal detectable systems
  • Lightweight design
  • Less costly alternative to stainless steel
Catalog No. 50-283-8411 Supplier MSC Supplier No. 53375556
This metal detectable mini bench scraper provides an excellent solution for bench top scraping needs. Its semi-ferrous additive allows for use with metal detectable systems. This plastic scraper offers a lightweight and less costly alternative to stainless steel.
  • Metal detectable
  • Semi-ferrous additive for use with metal detectable systems
  • Lightweight design
  • Less costly alternative to stainless steel