Learn More
Aviva Systems Biology Corp TESMIN Antibody - N-terminal region (ARP53623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MTL5. Alternate names: CXCDC2, MTLT, TESMINApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-61073