Learn More
Aviva Systems Biology Corp Mtm1 Antibody - N-terminal region (ARP56078_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56078_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mtm1. It was validated on Western Blot. . Alternate names: AF073996, KIAA4176, Mtm, mKIAA4176ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-114