Learn More
Aviva Systems Biology Corp MTMR12 Antibody - middle region (ARP56273_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56273_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTMR12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MTMR12. Alternate names: 3-PAP, PIP3APApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61790