Learn More
Aviva Systems Biology Corp MTMR14 Antibody - middle region (ARP57637_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57637_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTMR14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MTMR14. Alternate names: C3orf29, FLJ22405, FLJ90311, hEDTPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62875