Learn More
ABclonal Technology MTMR4 Rabbit pAb
SDP
Catalog No. 5000108154
Encompass_Preferred
Enables protein phosphatase binding activity. Involved in regulation of phosphatidylinositol dephosphorylation. Located in endosome.
Catalog No. 50-001-08154 Supplier ABclonal Technology Supplier No. A15349500UL
Enables protein phosphatase binding activity. Involved in regulation of phosphatidylinositol dephosphorylation. Located in endosome.