Learn More
ABclonal Technology MTMR6 Rabbit pAb
SDP
Catalog No. 5000106973
Encompass_Preferred
Enables phosphatidylinositol-3,5-bisphosphate phosphatase activity and phosphatidylinositol-3-phosphatase activity. Involved in phosphatidylinositol dephosphorylation. Located in cytoplasm and nuclear envelope.
Catalog No. 50-001-06973 Supplier ABclonal Technology Supplier No. A153481000UL
Enables phosphatidylinositol-3,5-bisphosphate phosphatase activity and phosphatidylinositol-3-phosphatase activity. Involved in phosphatidylinositol dephosphorylation. Located in cytoplasm and nuclear envelope.