Learn More
Eurofins Discoverx PathHunter MTNR1A Total Internalization DLD1 Cell Line, 2 vials. (93-0847C13)
Supplier:  Eurofins Discoverx 930847C13
Encompass
The PathHunter MTNR1A Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of MTNR1A via detection of localization to the early endosome. The included cell line overexpresses EA-tagged MTNR1A and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the MTNR1A receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1416
Add to Cart
Request a Quote