Learn More
Aviva Systems Biology Corp MTO1 Antibody - N-terminal region (ARP54841_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54841_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MTO1. Alternate names: CGI-02ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61338