Learn More
Medchemexpress LLC HY-112914 5mg Medchemexpress, mTOR inhibitor-1 CAS:468747-17-3 Purity:>98%
SDP
Catalog No. 501932444
Encompass_Preferred
Medchemexpress, HY-112914 5mg mTOR inhibitor-1 CAS:468747-17-3 mTOR inhibitor-1 is a novel mTOR pathway inhibitor which can suppress cells proliferation and inducing autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2444 Supplier Medchemexpress LLC Supplier No. HY1129145MG
Medchemexpress, HY-112914 5mg mTOR inhibitor-1 CAS:468747-17-3 mTOR inhibitor-1 is a novel mTOR pathway inhibitor which can suppress cells proliferation and inducing autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.