Learn More
TARGETMOL CHEMICALS INC MTOR INHIBITOR-3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T121235MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. mTOR inhibitor-3 is a selective mTOR inhibitor (Ki= 1.5 nM). mTOR inhibitor-3 inhibits mTORC1 and mTORC2 in cellular and in vivo experiments. purity: 98%
Catalog No. 50-258-1482