Learn More
Medchemexpress LLC mTOR inhibitor-8 | 2489196-70-3 | 98.1% | C24H19ClN4OS | 10MG
SDP
Catalog No. 5000260873
Encompass_Preferred
mTOR inhibitor-8 is a small-molecule inhibitor of the mechanistic target of rapamycin (mTOR) that acts via FKBP12 to inhibit mTOR activity and induces autophagy in A549 human lung cancer cells. It is provided as a solid research reagent and is available in multiple sizes and solution formats for use in cell-based assays.
  • Inhibits mTOR activity via FKBP12.
  • Induces autophagy in A549 human lung cancer cells.
  • Supplied as a solid and as a 10 mM solution in DMSO for assay flexibility.
  • High purity (98.1%) suitable for research applications.
  • Defined chemical formula (C24H19ClN4OS) and molecular weight for accurate identification.
Catalog No. 50-002-60873 Supplier Medchemexpress LLC Supplier No. HY13134410MG
Add to Cart
Add to Cart
mTOR inhibitor-8 is a small-molecule inhibitor of the mechanistic target of rapamycin (mTOR) that acts via FKBP12 to inhibit mTOR activity and induces autophagy in A549 human lung cancer cells. It is provided as a solid research reagent and is available in multiple sizes and solution formats for use in cell-based assays.
  • Inhibits mTOR activity via FKBP12.
  • Induces autophagy in A549 human lung cancer cells.
  • Supplied as a solid and as a 10 mM solution in DMSO for assay flexibility.
  • High purity (98.1%) suitable for research applications.
  • Defined chemical formula (C24H19ClN4OS) and molecular weight for accurate identification.