Learn More
Medchemexpress LLC HY-131344 5mg Medchemexpress, mTOR inhibitor-8 CAS:2489196-70-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1313445MG
Encompass_Preferred
Medchemexpress, HY-131344 5mg mTOR inhibitor-8 CAS:2489196-70-3 mTOR inhibitor-8 is an mTOR inhibitor and autophagy inducer. mTOR inhibitor-8 inhibits the activity of mTOR via FKBP12 and induces autophagy of A549 human lung cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1278