Learn More
TARGETMOL CHEMICALS INC MTOR INHIBITOR 9B 5MG
SDP
Catalog No. 502579282
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. mTOR inhibitor 9b is an enzyme inhibitor of protein kinase mTOR he phosphatidylinositol 3-kinase PIK3CA with IC50s of 0.76 nm and 1.262 µM respectively.mTOR inhibitor 9b possesses anticancer activity and can be used to study leukemia skin breast lung and colon cancers. purity: 99%
Catalog No. 50-257-9282 Supplier TARGETMOL CHEMICALS INC Supplier No. T677035MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. mTOR inhibitor 9b is an enzyme inhibitor of protein kinase mTOR he phosphatidylinositol 3-kinase PIK3CA with IC50s of 0.76 nm and 1.262 µM respectively.mTOR inhibitor 9b possesses anticancer activity and can be used to study leukemia skin breast lung and colon cancers. purity: 99%