Learn More
Enzo Life Sciences mTOR inhibitor WYE-23 (1mg). CAS: 1062169-46-3
SDP
Catalog No. 502013431
Encompass_Preferred
Alternative name: 4-{6-[4-(3-Cyclopropyl-ureido)-phenyl]-4-morpholin-4-yl-pyrazolo[3,4-d]pyrimidin-1-yl}-piperidine-1-carboxylic acid methyl ester. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3431 Supplier Enzo Life Sciences Supplier No. ENZCHM1360001
Alternative name: 4-{6-[4-(3-Cyclopropyl-ureido)-phenyl]-4-morpholin-4-yl-pyrazolo[3,4-d]pyrimidin-1-yl}-piperidine-1-carboxylic acid methyl ester. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.