Learn More
Enzo Life Sciences mTOR inhibitor WYE-28 (1mg). CAS: 1062172-60-4
SDP
Catalog No. 502013433
Encompass_Preferred
Cell-permeable. A potent, selective, and ATP-competitive mTOR (mammalian target of rapamycin) inhibitor (IC50 = 0.08 nM). WYE-28 displays 75-fold selectivity over PI3Kα (IC50 = 6 nM). Alternative name: 4-(6-{4-[3-(4-Hydroxymethyl-phenyl)-ureido]-phenyl}-4-morpholin-4-yl-pyrazolo[3,4-d]pyrimidin-1-yl)-piperidine-1-carboxylic acid methyl ester. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3433 Supplier Enzo Life Sciences Supplier No. ENZCHM1370001
Cell-permeable. A potent, selective, and ATP-competitive mTOR (mammalian target of rapamycin) inhibitor (IC50 = 0.08 nM). WYE-28 displays 75-fold selectivity over PI3Kα (IC50 = 6 nM). Alternative name: 4-(6-{4-[3-(4-Hydroxymethyl-phenyl)-ureido]-phenyl}-4-morpholin-4-yl-pyrazolo[3,4-d]pyrimidin-1-yl)-piperidine-1-carboxylic acid methyl ester. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.