Learn More
STEMCELL Technologies Torin 2, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 1000260
Encompass
Torin 2 is a potent and selective ATP-competitive inhibitor against mTOR (EC₅₀ = 0.25 nM), a key regulator of cell growth, survival, and autophagy (Liu et al.; Petherick et al.). CANCER RESEARCH· Inhibits proliferation of lung, breast, colorectal, and cervical cancer cell lines (Liu et al.).· Blocks mTOR complex 1 (mTORC1)-associated cell cycle progression and induces autophagy in hepatocellular carcinoma cells (Wang et al.).
Catalog No. 50-204-5504