Learn More
Raybiotech Inc mTOR Pathway Screening Array, 4 Sample Kit
Catalog No. NC3888367
Encompass_Preferred
Screen the relative levels of 118 human proteins involved in mTOR signaling pathway. Suitable for serum, plasma, cell culture supernatants, and lysates.
Catalog No. NC3888367 Supplier Raybiotech Inc Supplier No. AAHBLGTOR4
Screen the relative levels of 118 human proteins involved in mTOR signaling pathway. Suitable for serum, plasma, cell culture supernatants, and lysates.